Cold storage overalls with bib and braces, Zermatt 6105
Supplier: SIOEN
Certificates
About this item
Protective overalls made of polyester and cotton, with a fixed quilted lining inside. The trousers provide protection from temperatures up to −40 °C and avoid cold bridges.
- Extra protection and lining at knees
- High back, stitched seams
- Zip closure on the back of legs
- Two inset pockets
- Braces with buckles
- Ankles adjustable by touch and close fastening, zip closure under flap at leg
Weight: 225 g/m²
Certifications: ENV 342 (3-2)
Specifications
- Catalog No:
- 113-6057
- 113-6056
- 113-6055
- 113-6054
- 113-6059
- 113-6058
- Modello:
- Zermatt 6105
- Zermatt 6105
- Zermatt 6105
- Zermatt 6105
- Zermatt 6105
- Zermatt 6105
- Applicazioni per abiti:
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Risk area:
- Cryo, Cold
- Cryo, Cold
- Cryo, Cold
- Cryo, Cold
- Cryo, Cold
- Cryo, Cold
- Genere:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Tipo di chiusura:
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Tessuto:
- polyester and cotton
- polyester and cotton
- polyester and cotton
- polyester and cotton
- polyester and cotton
- polyester and cotton
- Peso:
- 225 g/m²
- 225 g/m²
- 225 g/m²
- 225 g/m²
- 225 g/m²
- 225 g/m²