T-shirts, A-Code® Basecamp
Supplier: FRISTADS KANSAS
Certificates
About this item
Tubular knit T-shirts made from 100% ring-spun cotton (single jersey). The grey melange shirt is made from 85% cotton and 15% viscose, while the yellow version is made from 65% polyester and 35% cotton.
- Ribbed cuffs with elastane content
- Crew neck
- Cotton is pre-shrunk to prevent shrinking during washing
Weight: 190 g/m²
Specifications
- Catalog No:
- 113-2663
- 113-2647
- 113-2664
- 113-2648
- 113-2665
- 113-2666
- 113-2643
- 113-2661
- 113-2668
- 113-2669
- 113-2670
- 113-2619
- 113-2660
- 113-2655
- 113-2604
- 113-2605
- 113-2606
- 113-2597
- 113-2622
- 113-2644
- 113-2623
- 113-2645
- 113-2662
- 113-2624
- 113-2646
- 113-2674
- 113-2675
- 113-2676
- 113-2677
- 113-2678
- 113-2673
- 113-2633
- 113-2636
- 113-2659
- 113-2598
- 113-2599
- 113-2620
- 113-2621
- 113-2671
- 113-2672
- 113-2667
- 113-2079
- 113-2082
- 113-2617
- 113-2618
- 113-2075
- 113-2656
- 113-2657
- 113-2658
- Applicazioni per abiti:
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Genere:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Tessuto:
- 100% Cotton
- 85% cotton, 15% viscose
- 100% Cotton
- 85% cotton, 15% viscose
- 100% Cotton
- 100% Cotton
- 85% cotton, 15% viscose
- 100% Cotton
- 100% Cotton
- 100% Cotton
- 100% Cotton
- 100% Cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 100% Cotton
- 100% Cotton
- 100% Cotton
- 100% Cotton
- 100% Cotton
- 85% cotton, 15% viscose
- 100% Cotton
- 85% cotton, 15% viscose
- 100% Cotton
- 100% Cotton
- 85% cotton, 15% viscose
- 100% Cotton
- 100% Cotton
- 100% Cotton
- 100% Cotton
- 100% Cotton
- 100% Cotton
- 100% Cotton
- 100% Cotton
- 65% polyester, 35% cotton
- 100% Cotton
- 100% Cotton
- 100% Cotton
- 100% Cotton
- 100% Cotton
- 100% Cotton
- 100% Cotton
- 100% Cotton
- 100% Cotton
- 100% Cotton
- 100% Cotton
- 100% Cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- 65% polyester, 35% cotton
- Peso:
- 190 g/m²
- 190 g/m²
- 190 g/m²
- 190 g/m²
- 190 g/m²
- 190 g/m²
- 190 g/m²
- 190 g/m²
- 190 g/m²
- 190 g/m²
- 190 g/m²
- 190 g/m²
- 190 g/m²
- 190 g/m²
- 190 g/m²
- 190 g/m²
- 190 g/m²
- 190 g/m²
- 190 g/m²
- 190 g/m²
- 190 g/m²
- 190 g/m²
- 190 g/m²
- 190 g/m²
- 190 g/m²
- 190 g/m²
- 190 g/m²
- 190 g/m²
- 190 g/m²
- 190 g/m²
- 190 g/m²
- 190 g/m²
- 190 g/m²
- 190 g/m²
- 190 g/m²
- 190 g/m²
- 190 g/m²
- 190 g/m²
- 190 g/m²
- 190 g/m²
- 190 g/m²
- 190 g/m²
- 190 g/m²
- 190 g/m²
- 190 g/m²
- 190 g/m²
- 190 g/m²
- 190 g/m²
- 190 g/m²