High visibility overalls with bib and braces, Amberg 6623
Supplier: SIOEN
Certificates
About this item
Bib and brace trousers made of Flexothane® Essential (100% polyester knitting with PU coating), waterproof and lightweight.
- Bib in front, elastic braces with PVC buttons and buckles
- High frequency welded seams
- Front fly with press studs
- Legs adjustable by press studs
- 3M™ reflective tape
Weight: 170 g/m²
Certifications: EN 471 (2-2), ENV 343 (3-1)
Specifications
- Catalog No:
- 113-07160
- 113-07157
- 113-07158
- 113-07159
- 113-07162
- 113-07161
- Modello:
- Amberg Flexothane Essential
- Amberg Flexothane Essential
- Amberg Flexothane Essential
- Amberg Flexothane Essential
- Amberg Flexothane Essential
- Amberg Flexothane Essential
- Applicazioni per abiti:
- General Purpose, Industrial
- General Purpose, Industrial
- General Purpose, Industrial
- General Purpose, Industrial
- General Purpose, Industrial
- General Purpose, Industrial
- Risk area:
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis
- Genere:
- Per l'uomo
- Per l'uomo
- Per l'uomo
- Per l'uomo
- Per l'uomo
- Per l'uomo
- Sleeve or leg length:
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Tipo di chiusura:
- Con bottoni
- Con bottoni
- Con bottoni
- Con bottoni
- Con bottoni
- Con bottoni
- Tessuto:
- 100% polyester knitting with PU coating
- 100% polyester knitting with PU coating
- 100% polyester knitting with PU coating
- 100% polyester knitting with PU coating
- 100% polyester knitting with PU coating
- 100% polyester knitting with PU coating
- Peso:
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²