High visibility rain jacket, Kassel 4279
Supplier: SIOEN
Certificates
About this item
Waterproof and windproof jacket made of Flexothane® Essential (100% polyester knitting with PU coating). With high frequency welded seams and rain gutter.
- With 3M™ reflective tape
- Straight collar, foldaway hood in collar, zip closure under flap with press studs
- Two inset pockets
- Raglan sleeves, sleeve narrowing by press studs
- Back ventilation
Suitable for zipping in linings or fleeces.
Back length: 88 cm
Weight: 170 g/m²
Certifications: EN 471 (3-2), ENV 343 (3-1)
Specifications
- Catalog No:
- 113-5940
- 113-5950
- 113-5941
- 113-5947
- 113-5949
- 113-5946
- 113-5948
- 113-5942
- 113-5943
- 113-5944
- 113-5945
- 113-5939
- Modello:
- 4279 Kassel
- 4279 Kassel
- 4279 Kassel
- 4279 Kassel
- 4279 Kassel
- 4279 Kassel
- 4279 Kassel
- 4279 Kassel
- 4279 Kassel
- 4279 Kassel
- 4279 Kassel
- 4279 Kassel
- Applicazioni per abiti:
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Risk area:
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Genere:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Tipo di chiusura:
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Tessuto:
- Flexothane® Essential (100% polyester knitting with PU coating)
- Flexothane® Essential (100% polyester knitting with PU coating)
- Flexothane® Essential (100% polyester knitting with PU coating)
- Flexothane® Essential (100% polyester knitting with PU coating)
- Flexothane® Essential (100% polyester knitting with PU coating)
- Flexothane® Essential (100% polyester knitting with PU coating)
- Flexothane® Essential (100% polyester knitting with PU coating)
- Flexothane® Essential (100% polyester knitting with PU coating)
- Flexothane® Essential (100% polyester knitting with PU coating)
- Flexothane® Essential (100% polyester knitting with PU coating)
- Flexothane® Essential (100% polyester knitting with PU coating)
- Flexothane® Essential (100% polyester knitting with PU coating)
- Peso:
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²
- 170 g/m²