Welding overalls, Flam
Supplier: FRISTADS KANSAS
Certificates
About this item
Protective overalls made from Flam fabric (75% cotton, 25% polyester), which offers long lasting protection against open flames and sources of heat thanks to the flame-retardant used.
- Concealed two-way zip and buttons up to the top of the collar
- Two breast pockets with flap, one with pen loops; inner pocket with mobile phone pouch and loop for earphones, two front pockets
- Back pockets with flap and concealed press studs, leg pocket with flap and concealed press studs, folding-rule pocket with extra pocket
- Pre-formed knee with knee pockets
- Adjustable cuffs with rip-tape fastening, adjustable waist
The overalls are ideal for tasks where welding sparks, open flames or even arcing faults can threaten clothing, and thus pose a risk to the wearer.
Weight: 340 g/m²
Certifications: EN 61482-1-2 (class 1), EN ISO 11611 (A1 class 1), EN ISO 11612 (A1 B1 C1)
Specifications
- Catalog No:
- 113-3914
- 113-3918
- 113-3915
- 113-3917
- 113-3916
- Modello:
- FLAM
- FLAM
- FLAM
- FLAM
- FLAM
- Applicazioni per abiti:
- Welding
- Welding
- Welding
- Welding
- Welding
- Risk area:
- Heat, Flame-retardant
- Heat, Flame-retardant
- Heat, Flame-retardant
- Heat, Flame-retardant
- Heat, Flame-retardant
- Genere:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Tipo di chiusura:
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Tessuto:
- 75% cotton, 25% polyester
- 75% cotton, 25% polyester
- 75% cotton, 25% polyester
- 75% cotton, 25% polyester
- 75% cotton, 25% polyester
- Peso:
- 340 g/m²
- 340 g/m²
- 340 g/m²
- 340 g/m²
- 340 g/m²