Centrifuge tubes, ASTM, BLAUBRAND®, DAkks
BRNDDAKKS362038
: Brand
- Tipo di chiusura:-
- Capacità:100 ml
- Descrizione:Cylindrical, conical at the bottom, ASTM D 91
- Lunghezza:203 mm
- Conf:1
Borosilicate glass 3.3, graduated.
- Withstands forces up to 700×g
- Design, accuracy, etc. to meet ASTM specifications
- With DAkks-Certifikate