Surgical gloves, polyisoprene, sterile, Vasco® OP free
: B. Braun
These anatomically shaped gloves are designed to meet highest requirements in the operating theatre and in aseptic working procedures. Special washing procedures and a net-like inner lining structure allow very easy donning in single and double gloving. The ergonomic design guarantees perfect fit and high comfort, hand fatigue is avoided.
- Powder-free, micro-textured finish
- Latex-free inner polymer coating for easy donning
- Beaded conical cuff provides a safe border to the gown and well adjusted fit on the wrist
- Outstanding dry and wet grip performance and high acceptance in surgery
- Feels like a second skin, the sensitivity is not even compromised by double gloving
- Sensitising substances are reduced to a minimum by sophisticated surface treatment, resulting in excellent skin tolerance
Length: 270 - 285 mm
Thickness (single wall): 0,26 mm (finger), 0,19 mm (palm), 0,175 mm (cuff)
Colour: Beige
EN 455 (1-4), EN 374-3; AQL 1,0
: EN 455, EN ISO 374-1 Type B (KPTMS), EN ISO 374-5 VIRUS, EN 421; Cat. III; AQL 0,65
: One pair with folded cuffs packaged in sterile peel packs. 40 pairs per box.
- Catalog No:
- BRAU9208356
- BRAU9208330
- BRAU9208313
- Ambidestro/mano specifico:
- mano specifico
- mano specifico
- mano specifico
- Monouso/riutilizzabile:
- monouso
- monouso
- monouso
- Lattice/senza lattice:
- Senza lattice
- Senza lattice
- Senza lattice
- Talcati/non talcati:
- Non talcati
- Non talcati
- Non talcati
- Finitura:
- Micro-textured
- Micro-textured
- Micro-textured
- Tipo polsino:
- Arrotolato
- Arrotolato
- Arrotolato
- Spessore polsino:
- 0,175 mm
- 0,175 mm
- 0,175 mm
- Spessore delle dita:
- 0,185 mm
- 0,185 mm
- 0,185 mm
- Spessore palmo:
- 0,19 mm
- 0,19 mm
- 0,19 mm
- Norma EN:
- EN 455, EN ISO 374-1 Type B (KPTMS), EN ISO 374-5 VIRUS, EN 421
- EN 455, EN ISO 374-1 Type B (KPTMS), EN ISO 374-5 VIRUS, EN 421
- EN 455, EN ISO 374-1 Type B (KPTMS), EN ISO 374-5 VIRUS, EN 421
Frequently Bought Together