Bags, safety pouch, SP Bel-Art
Codice catalogo BELAH132340000
Nome del fornitore: Bel-Art Products, a Part of SP
Specifiche
- Confezione:200 pcs/pack
- Descrizione:Biohazard sharp object safety pouch, thickness 0,25 mm
- L×L:140×330 mm
- Materiale:Paperboard
- Conf:200
Specifiche
Informazioni su questo articolo
Hard to pierce pouch offering protection against the danger of being cut by contaminated sharp objects. Convenient for labs, clinics and medical offices. Used with the Poxygrid® safety pouch stand for easy benchtop collection.
- Solid paperboard pouch
- Remains open in an oval shape by folding the bottom flaps
- After filling, the top flaps close and lock in place for safe disposal
- Printed with bold biohazard symbol