Flow indicator, SP Bel-Art
PC, 6,4 - 9,5 mm, Flow indicator, 100 - 1000 ml/min
Catalog # BELAH199350000
Supplier: Bel-Art Products, a Part of SP
Jump to:
Product Details & Documents
Flow indicator with polypropylene non returning valve.
Specifications
- Massima velocità del flusso:100 - 1000 ml/min
- Descrizione:Flow indicator
- Materiale:PC
- Ø tubo:6,4 - 9,5 mm