Orders LinkContactUsLinkComponent
Buffer, TRIS-glycine solution 10X concentrate (TG buffer)
Buffer, TRIS-glycine solution 10X concentrate (TG buffer)
TRIS-glycine buffer solution 10X concentrate (TG buffer), bottiglia in vetro, 1 L
Codice catalogo NADIEC-880-1L
Marketsource
Nome del fornitore:   NATIONAL DIAGNOSTICS
Buffer, TRIS-glycine solution 10X concentrate (TG buffer)
Codice catalogo NADIEC-880-1L
Nome del fornitore:  NATIONAL DIAGNOSTICS
Marketsource
Numero CAS:  
Buffer, TRIS-glycine solution 10X concentrate (TG buffer)
TRIS-glycine buffer solution 10X concentrate (TG buffer), bottiglia in vetro, 1 L
Numero CAS:  
Price
Richiedi prezzo
Log in per disponibilità
1 * 1 L
Product Details & Documents
Specifiche
Specifiche
  • Tipo di conf.:
    bottiglia in vetro
  • Conf:
    1 L
  • Descrizione:
    TRIS-glycine buffer solution 10X concentrate (TG buffer)