Buffer, TE solution (TRIS-EDTA buffer) pH 8,0
TE buffer solution (TRIS-EDTA buffer) pH 8,0, bottiglia in vetro, 1 L
Catalog # MOLEM40814247
Marketsource
Supplier: Molekula
Buffer, TE solution (TRIS-EDTA buffer) pH 8,0
Catalog # MOLEM40814247
Supplier: Molekula
Marketsource
CAS Number:
Jump to:
Product Details & Documents
Specifications
Specifications
- Tipo di conf.:bottiglia in vetro
- Conf:1 L
- Descrizione:TE buffer solution (TRIS-EDTA buffer) pH 8,0