Buffer, SSC solution 20X concentrate (saline-sodium citrate)
SSC buffer solution 20X concentrate (saline-sodium citrate), 1 KIT
Catalog # EURLEMR255005BK
Supplier: EUROCLONE
Buffer, SSC solution 20X concentrate (saline-sodium citrate)
Catalog # EURLEMR255005BK
Supplier: EUROCLONE
CAS Number:
Jump to:
Product Details & Documents
Specifications
Specifications
- Conf:1 KIT
- Descrizione:SSC buffer solution 20X concentrate (saline-sodium citrate)
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



