Crucible tongs, titanium, with ridges
Catalog # BOCH2910
Supplier: BOCHEM
Specifications
- Lunghezza:220 mm
- Conf:1
- Capacità:20 mm
Specifications
About this item
Titanium crucible tong with ridges.
- Titanium is resistant to diluted acids and alkaline solutions
- Thermal resistant