ContactUsLinkComponent
Skimmed milk butyrometer, without stopper
Skimmed milk butyrometer, without stopper
Catalog # 614-0097
Supplier:  GERBER FUNKE
undefined
Skimmed milk butyrometer, without stopper
Catalog # 614-0097
Supplier:  GERBER FUNKE
Specifications
  • Descrizione:
    Butyrometer for skimmed milk, 0 - 1 % : 0,01, with open bulb
  • Conf:
    1
Specifications
About this item
According to Sichler`s method, with rounded scale.