Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 SFr.
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Menthol (non-specified)
Menthol (non-specified)
Menthol (non-specified)
ROTH6725.1
:
Roth Carl
:
Menthol (non-specified)
ROTH6725.1
:
Roth Carl
:
Tipo di conf.:
Glass bottle for solids
Conf:
250 g
Seq:
3545
:
:
156.265 g/mol
:
Ambiente
:
216-074-4
:
1490-04-6
:
C₁₀H₂₀O
:
MFCD00001484