Glycine ethyl ester hydrochloride (H-Gly-OEt.HCl)
Catalog # BACE4030677.0250
Supplier: BACHEM BIOCHEMICA
CAS Number:
Specifications
- Tipo di conf.:Bottiglia di vetro
- Conf:250 g
- Seq:3545
Specifications
About this item
:
:139.58 g/mol
EINECS:210-787-4
CAS Number:623-33-6
Formula:C₄H₁₀ClNO₂
MDL Number:MFCD00012871