Orders LinkContactUsLinkComponent
Milk butyrometer
Milk butyrometer
Supplier:   GERBER FUNKE
Milk butyrometer
Supplier:  GERBER FUNKE
Certificates
Milk butyrometer
16 Options Available Below
Product Details & Documents
Borosilicate glass.
  • With or without stopper
Available with a declaration of conformity per batch and type (in German only).
Product Family Options
Butyrometer, without stopper
Product Information
  • Descrizione
    Availability
    Price
  • Butyrometer for milk, 0 - 6% : 0,1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 4.40
    SFr. 4.40
    1 * 1 ST
       SFr. 4.40
  • Butyrometer for milk, 0 - 8% : 0,1
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - SFr. 27.00
    SFr. 27.00
    1 * 10 ST
       SFr. 27.00
Butyrometer, with stopper
Product Information
  • Descrizione
    Availability
    Price
  • Butyrometer for milk, 0 - 5% : 0,1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 16.70
    SFr. 16.70
    1 * 1 ST
       SFr. 16.70
  • Butyrometer for milk, 0 - 6% : 0,1
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - SFr. 35.40
    SFr. 35.40
    1 * 10 ST
       SFr. 35.40
  • Butyrometer for milk, 0 - 7% : 0,1
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - SFr. 35.40
    SFr. 35.40
    1 * 10 ST
       SFr. 35.40
  • Butyrometer for milk, 0 - 8% : 0,1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 4.60
    SFr. 4.60
    1 * 1 ST
       SFr. 4.60
  • Butyrometer for milk, 0 - 9% : 0,1
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - SFr. 113.00
    SFr. 113.00
    1 * 10 ST
       SFr. 113.00
  • Butyrometer for milk, 0 - 10% : 0,1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 4.60
    SFr. 4.60
    1 * 1 ST
       SFr. 4.60
  • Butyrometer for milk, 0 - 12% : 0,1
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - SFr. 114.00
    SFr. 114.00
    1 * 10 ST
       SFr. 114.00
  • Butyrometer for milk, 0 - 16% : 0,2
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - SFr. 120.00
    SFr. 120.00
    1 * 10 ST
       SFr. 120.00
Butyrometer, without stopper, with conformtity certificate
Product Information
  • Descrizione
    Availability
    Price
  • Butyrometer for milk, 0 - 5% : 0,1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 27.20
    SFr. 27.20
    1 * 1 ST
       SFr. 27.20
  • Butyrometer for milk, 0 - 9% : 0,1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 27.20
    SFr. 27.20
    1 * 1 ST
       SFr. 27.20
  • Butyrometer for milk, 0 - 12% : 0,1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 27.60
    SFr. 27.60
    1 * 1 ST
       SFr. 27.60
Butyrometer, with stopper, with conformtity certificate
Product Information
  • Descrizione
    Availability
    Price
  • Butyrometer for milk, 0 - 5% : 0,1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 27.80
    SFr. 27.80
    1 * 1 ST
       SFr. 27.80
  • Butyrometer for milk, 0 - 9% : 0,1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 27.80
    SFr. 27.80
    1 * 1 ST
       SFr. 27.80
  • Butyrometer for milk, 0 - 12% : 0,1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 28.20
    SFr. 28.20
    1 * 1 ST
       SFr. 28.20
Accessories
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • 614-2028
    Stopper, rubber, conical, for all measuring method butyrometers
    Log in for availability
    1 * 100 ST - SFr. 44.90
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.