Joining pieces for flat flanges, BOLA
Joining piece with 8 screws, silumin, Flange NW 170/146
Codice catalogo BOHLB323-01
Nome del fornitore: Bohlender
Vai a
Product Details & Documents
Made of aluminium.
- For flanges NW 170/146
Connection between reaction vessel and lid. Locked by zinc-plated steel screws.
Specifiche
- Per:Flange NW 170/146
- Descrizione:Joining piece with 8 screws, silumin