Sodium azide tablets for milk preservation, Sigma-Aldrich®
Catalog # 1.06687.0001
Anbieter: Merck
CAS-Nummer:
Technische Daten
- Conf:5.000 Tab.
- Descrizione:Sodium azide tablets
Technische Daten
Über diesen Artikel
Simple to use tablet for preservation of milk samples prior to testing. Each 100 mg tablet contains approximately 8 mg of sodium azide.
Packaging: Packed in plastic can.