SP Bel-Art sidENTRY™, Glove box
Catalog # BELAH500260000
Supplier: Bel-Art Products, a Part of SP
Specifications
- Descrizione:Camera a guanti, in acrilico, con due orifizi anteriori circolari per guanti (Ø 203,2 mm) e uno sportello laterale grande con guarnizione (44,5 cm²)
- L×P×A est.:762×610×610 mm
- Conf:1
Specifications
About this item
This small and portable glove box is designed for remote and field use. It is manufactured from transparent acrylic plastic (6,35 mm thick).
- Glove box stability is assured by four adjustable levelers that also level the box and all its contents
- Stopcocks are included ready for assembly
- Includes large gasketed door on side of unit
- Features two 20,32 cm glove ports compatible with standard glove box gloves