Substrate for the detection of immobilised HRP
Catalog # 732-1208
Supplier: Uptima
CAS Number:
Specifications
- Conf:1 KIT
- Descrizione:UptiLight
Specifications
About this item
Substrate optimised for the detection of immobilised HRP in Western, Southern, Northern and dot blot applications.
- Compatibility with nylon, nitrocellulose and PVDF membranes
- Fast response time - specific detection in 1 to 10 minutes
- High sensitivity - detects up to 1 to 2 pg antigen
- Long signal duration - stable reagent allows repeated exposure