Orders LinkContactUsLinkComponent
Drip closures | Avantor
Drip closures
Supplier:   MUELLER KREMPEL
Drip closures
Supplier:  MUELLER KREMPEL
Certificates
Drip closures
3 Options Available Below
Product Details & Documents
Product Family Options
Product Information
  • Descrizione
    Availability
    Price
  • Vertical dropper for aqueous solutions; White tamper evident closure with dropper insert for openings according to DIN-18 for low surface tension aqueous solutions; also containing ethanol up to 15%
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - SFr. 26.50
    SFr. 26.50
    1 * 100 ST
       SFr. 26.50
  • Vertical dropper for thinly flowing, essential oils; Tamper evident closure with dropper insert for openings according to DIN-18
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - SFr. 26.50
    SFr. 26.50
    1 * 100 ST
       SFr. 26.50
  • Vertical dropper for medium viscosity oils; Tamper evident closure with dropper insert for openings according to DIN-18, for medium viscosity oils
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - SFr. 26.50
    SFr. 26.50
    1 * 100 ST
       SFr. 26.50