Scalpel blade remover unit
Scalpel blade remover unit, single unit
Nome del fornitore: SWANN MORTON
Vai a
Product Details & Documents
Plastic box designed for the removal of blades, not for long term sharps storage.
- Holds up to 100 blades
- Very useful in areas where a sharps disposal facility is not immediately available
Specifiche
- Descrizione:Scalpel blade remover unit, single unit