Staining jars according to Coplin, WHEATON®
Staining jar acc. to Coplin, with PP screw cap, 26×26×90 mm
Codice catalogo 631-2509
Nome del fornitore: DWK Life Sciences
Staining jars according to Coplin, WHEATON®
Codice catalogo 631-2509
Nome del fornitore: DWK Life Sciences
Vai a
Product Details & Documents
Soda-lime glass.
- Linerless, white PP screw cap to reduce solvent evaporation
- Capacity: Approx. 60 ml
This unit is used for staining slides, or as a developing chamber for thin layer chromatography. It holds five single 76×26 mm slides vertically, or 10 slides back to back.
Specifiche
- Descrizione:Staining jar acc. to Coplin, with PP screw cap
- L×P×A:26×26×90 mm
Accessories
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Slide grip for Wheaton Coplin staining jars