Complete sampler, Wheaton® E-Z
Complete sampler, PP, 125 ml
Codice catalogo WHEA885020
Nome del fornitore: DWK Life Sciences
Vai a
Product Details & Documents
The filling ports are located above the bottle to ensure that it is completely filled to reduce the possibility of air entrapment.
- Head/shaft assembly and borosilicate glass bottle with lined cap
- Constructed of chemically resistant PP
- Bottle with a GPI 33-430 thread
This sampler is an efficient and time saving method of sampling from drums and tanks.
Specifiche
- Capacità:125 ml
- Descrizione:Complete sampler
- Materiale:PP
Accessories
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Flacone, 125 ml, tappo con liner