Large insulating dewar flasks, type 131 - 135
About this item
Dewar flasks are made of glass and are reliable and economical with a protective casing out of blue-coated metal. These are used in laboratory techniques, medicinal techniques, biotechnology, storing of CO₂ and pressure-less space for coolants.
- With side grips
- Lid made of metal, blue coated with insulating disc
- Lid with over pressure valve
- Lockable lid and roller base
Certifications: Glass material according to ISO 3585 and Dewar flasks according to DIN 12492.
2 Options Available Below
Product Details & Documents
Dewar flasks are made of glass and are reliable and economical with a protective casing out of blue-coated metal. These are used in laboratory techniques, medicinal techniques, biotechnology, storing of CO₂ and pressure-less space for coolants.
- With side grips
- Lid made of metal, blue coated with insulating disc
- Lid with over pressure valve
- Lockable lid and roller base
Certifications: Glass material according to ISO 3585 and Dewar flasks according to DIN 12492.
Specifications for Product Family
Specifications
- Catalog No:
- KGWA1263
- KGWA1265
- Modello:
- 132
- 134
- Capacità:
- 14 L
- 28 L
- Evaporazione giornaliera:
- 0,4 L
- 0,6 L
- Altezza totale:
- 635 mm
- 765 mm
Specifications
Product Family Options
Product Information
- DescrizioneAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.KGWA1263Modello132Capacità14 LEvaporazione giornaliera0,4 LAltezza totale635 mm - Specifications:
More
Specifications
Cat. No.KGWA1265Modello134Capacità28 LEvaporazione giornaliera0,6 LAltezza totale765 mm
Accessories
Product Information
- Catalog #DescrizioneAvailabilityPrice
Product Information
- Catalog #DescrizioneAvailabilityPrice
Product Information
- Catalog #DescrizioneAvailabilityPrice
Product Information
- Catalog #DescrizioneAvailabilityPrice
Product Information
- Catalog #DescrizioneAvailabilityPrice
Product Information
- Catalog #DescrizioneAvailabilityPrice
Product Information
- Catalog #DescrizioneAvailabilityPrice
Product Information
- Catalog #DescrizioneAvailabilityPrice
Product Information
- Catalog #DescrizioneAvailabilityPrice
Product Information
- Catalog #DescrizioneAvailabilityPrice