Cryogenic freezer boxes, CryoFile™, WHEATON®
High quality chip-board, spray coated with polyethylene.
- Water repellent material allows for longer durability
- Numbering system printed on lid and bottom of the box
- Withstands cryogenic temperatures to –196 °C
- Six colours provide easy sample idenfication
Use for batching and storing cryogenic vials or ampoules. Partitions numbered from 1 to 81 for easy content identification.
High quality chip-board, spray coated with polyethylene.
- Water repellent material allows for longer durability
- Numbering system printed on lid and bottom of the box
- Withstands cryogenic temperatures to –196 °C
- Six colours provide easy sample idenfication
Use for batching and storing cryogenic vials or ampoules. Partitions numbered from 1 to 81 for easy content identification.
Documents
VWRBRO_007b Biobanking_EU
Documentation produit
Product Family Options
CryoFile™ boxes
- ScompartiPerColoreEsterno L×P×A
- 9×9Cryovials 1 - 2 mlverde130×130×52 mm1 * 15 STSFr. 218.00
- 9×9Cryovials 1 - 2 mlgiallo130×130×52 mm1 * 15 STSFr. 218.00
- 9×9Cryovials 1 - 2 mlrosa130×130×52 mm1 * 15 STSFr. 218.00
- 9×9Cryovials 1 - 2 mlbianco130×130×52 mm1 * 15 STSFr. 233.00
- 9×9Cryovials 1 - 2 mlblu130×130×52 mm1 * 15 STSFr. 233.00
- 9×9Cryovials 1 - 2 mlrosso130×130×52 mm1 * 15 STSFr. 218.00
CryoFile XL™ boxes
- ScompartiPerColoreEsterno L×P×A
- 9×9Cryovials 5 mlverde130×130×97 mm1 * 15 STSFr. 239.00
- 9×9Cryovials 5 mlgiallo130×130×97 mm1 * 15 STSFr. 239.00
- 9×9Cryovials 5 mlbianco130×130×97 mm1 * 15 STSFr. 239.00
- 9×9Cryovials 5 mlrosso130×130×97 mm1 * 15 STSFr. 239.00
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.