Orders LinkContactUsLinkComponent
Snap valves, BOLA
Snap valves, BOLA
Nome del fornitore:   Bohlender
Snap valves, BOLA
Nome del fornitore:  Bohlender
Certificates
Snap valves, BOLA
Snap valves, BOLA
2 varianti
Product Details & Documents
Body made of PTFE with 2 connections. Nuts made of glass-fibre reinforced PTFE for connecting tubing or tube.
  • Motionless sealing by means of a gastight bellow made of PTFE with flat cone point, no leakages
  • Temperature resistant upto 110 °C
  • Universal chemical resistance
  • Pressure upto 6 bar and suitable for vacuum
Snap valves suitable for panel mounting and easy disassembly for cleaning.
Lever locks when valve is open, switch setting is easily visible at position of the lever.
Product Family Options
Informazioni sul prodotto
  • Ø foro
    per tubo con Ø esterno
    Per
    Filettatura
    Disponibilità
    Price
  • 6 mm
    8 mm
    montaggio a pannello
    M 18×2
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 750.00
    SFr. 750.00
    1 * 1 ST
       SFr. 750.00
  • 6 mm
    10 mm
    montaggio a pannello
    M 18×2
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 750.00
    SFr. 750.00
    1 * 1 ST
       SFr. 750.00