Transfer spike for sterile liquids, Transofix®
Catalog # 720-2571
Supplier: B. Braun
Specifications
- Descrizione:Spike per liquidi sterili per flaconi fino a 100 ml
- Conf:50
Specifications
About this item
Transofix® Transfer Set is ideal for transferring sterile liquids.
- Conveniently designed large grip plates
- Free of latex and PVC
- Large internal diameter