Pipette guide, 'Guide 300'
Catalog # 613-1451
Supplier: DIVERSIFIED BIOTECH
Specifications
- Descrizione:Guida per pipette 'Guide 300'
- Conf:1
Specifications
About this item
This plastic guide enables users to easily target microplate wells using the plastic sliding grid.
- Reduces eyestrain during tedious pipetting work
- Suitable for use with multi channel pipettes
- Adjustable height to accommodate flat or v-bottom micro or PCR plates
The sliding guide is clearly marked with letters for fast and easy orientation. Ideal for long-term pipetting.