Orders LinkContactUsLinkComponent
Columns, TSK-Gel® NPH
Columns, TSK-Gel® NPH
Columns, TSK-Gel® NPH
Supplier:  TOSOH Bioscience
Certificates
Columns, TSK-Gel® NPH
Columns, TSK-Gel® NPH
5 Options Available Below
Product Details & Documents
Stainless steel columns on polymer basis.
Documents
Product Family Options
Guard columns
Product Information
  • Descrizione
    Formato della colonna
    Dimensioni particelle
    Ø int.
    Lunghezza
    Availability
    Price
  • DEAE-NPR
    Precolonna
    2,5 µm
    4,60 mm
    5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 656.00 SFr.
    656.00 SFr.
    1 * 1 ST
       656.00 SFr.
  • DNA-NPR
    Precolonna
    2,5 µm
    4,60 mm
    5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 795.00 SFr.
    795.00 SFr.
    1 * 1 ST
       795.00 SFr.
Product Information
  • Descrizione
    Formato della colonna
    Dimensioni particelle
    Ø int.
    Lunghezza
    Availability
    Price
  • SAX (IEX)
    5 µm
    6,00 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 970.00 SFr.
    1 970.00 SFr.
    1 * 1 ST
       1 970.00 SFr.
  • DEAE-NPR (IEX)
    2,5 µm
    4,60 mm
    35 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 540.00 SFr.
    1 540.00 SFr.
    1 * 1 ST
       1 540.00 SFr.
  • DNA-NPR (IEX)
    2,5 µm
    4,60 mm
    75 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 2 330.00 SFr.
    2 330.00 SFr.
    1 * 1 pezzi
       2 330.00 SFr.