Magnetic stirring bars, PTFE, Spinfin®, SP Bel-Art
Catalog # 442-9360
Supplier: Bel-Art Products, a Part of SP
Specifications
- Ø:7,9 mm
- Altezza:9,5 mm
- Seq:01
- Conf:1
Specifications
About this item
Spinfin® magnetic stirring bars agitate liquids in round bottom vessels such as test tubes, cylinders and flasks where other stirring bars cannot be used. Bars maintain their position in a vessel and will not wobble.