Stainless steel manipulator, Corning® HYPERStack®
Manipolatore HYPERStack, in acciaio inox
Codice catalogo 734-2491
Nome del fornitore: Corning
Stainless steel manipulator, Corning® HYPERStack®
Codice catalogo 734-2491
Nome del fornitore: Corning
Vai a
Product Details & Documents
The Corning® HYPERStack™ Stainless Steel Manipulator accommodates both HYPERStack™-36 and CellSTACK-10 vessels.
It handles two units of the same type simultaneously or one unit at a time. It is ergonomically designed and has large, easy to read dials that are clearly marked to identify positions for ease of use in protocol development.
Documents
Specifiche
- Descrizione:Manipolatore HYPERStack, in acciaio inox