Orders LinkContactUsLinkComponent
Supports for stand grids
Supports for stand grids
Supplier:   Lenz Laborglas
Supports for stand grids
Supplier:  Lenz Laborglas
Certificates
Supports for stand grids
Supports for stand grids
4 Options Available Below
Product Details & Documents
Stainless steel (clamp), steel, powder coated (supports).
For fixing lids onto reaction vessels and for the suspension into stands, complete with clamp and supports.
Product Family Options
Product Information
  • Per
    Availability
    Price
  • Flat flanges LF 100, framework grid 0,500 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 2’230.00
    SFr. 2’230.00
    1 * 1 ST
       SFr. 2’230.00
  • Flat flanges LF 150, framework grid 0,500 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 2’430.00
    SFr. 2’430.00
    1 * 1 ST
       SFr. 2’430.00
  • Flange piatte LF 200, struttura a griglia 0,500 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 3’320.00
    SFr. 3’320.00
    1 * 1 ST
       SFr. 3’320.00
  • Flange piatte LF 200, struttura a griglia 0,707 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 3’320.00
    SFr. 3’320.00
    1 * 1 ST
       SFr. 3’320.00