Storage containers, Tubby®
HEATHS23453
: HEATHROW
- L×P×A:330×200×115 mm
- Conf:5
Constructed of high-impact polypropylene, these sturdy storage tubs with lid and dividers are designed to help organize a laboratory.
- Removable dividers can create as big or as small a compartment as needed
- The lid snaps in place to secure the contents and for safe transport
- Perfect for keeping tubes, tips, pipettes, gloves, and other small items organized and out of the way