Cleanroom gloves, nitrile, sterile, BioClean N-Plus™
: Nitritex
These hand-specific nitrile gloves with beaded cuffs provide good resistance against chemicals and cytotoxic drugs. They also have excellent ESD properties. Suitable for chemotherapy applications.
- Sterile, powder-free, textured finish for enhanced grip
- Contain no natural latex proteins, hypoallergenic
- Extended length provides protection up to the elbow
- Beaded cuffs for easy donning
Suitable for use in Class 10, ISO 4 or M2.5 cleanrooms.
Typical particle count ≥0,5 μm: <1800 counts/cm²
Freedom from holes (final release): 1,0 AQL performance
EN 455, EN 374; Cat. III; AQL 1.5
: Manufactured in a facility holding ISO 9001:2015 and (EU)2016/425 Module D certifications.
Conforms to Category 3 - Complex Design Personal Protective Equipment PPE Regulation (EU)2016/425.
Processed in an NEBB certified ISO Class 4 environment.
Complies with the requirements of European Glove Standards EN 420:2003 +A1:2009, EN 374-1:2016 Type B, EN 374-5:2016 and EN 421:2010.
Physical properties comply with European Medical Glove Standard EN 455-2:2015.
Sterilized in accordance with ISO 11137-1:2006.
Suitable for contact with all foodstuffs. Regulation (EC)1935/2004 and (EC) 2023/2006.
Tested against permeation standard ASTM D6978-05 for cytotoxic drugs.
Conforms to Category 3 - Complex Design Personal Protective Equipment PPE Regulation (EU)2016/425.
Processed in an NEBB certified ISO Class 4 environment.
Complies with the requirements of European Glove Standards EN 420:2003 +A1:2009, EN 374-1:2016 Type B, EN 374-5:2016 and EN 421:2010.
Physical properties comply with European Medical Glove Standard EN 455-2:2015.
Sterilized in accordance with ISO 11137-1:2006.
Suitable for contact with all foodstuffs. Regulation (EC)1935/2004 and (EC) 2023/2006.
Tested against permeation standard ASTM D6978-05 for cytotoxic drugs.
: One cuffed pair per plastic wallet; one wallet per EasyTear™ PE pouch; 10 pairs per sealed outer PE bag; 20 outer bags per carton liner; one carton liner per carton (200 pairs)
- Catalog No:
- 112-3869
- 112-3871
- 112-3867
- 112-3866
- 112-3868
- 112-3865
- 112-3872
- 112-3870
- Ambidestro/mano specifico:
- mano specifico
- mano specifico
- mano specifico
- mano specifico
- mano specifico
- mano specifico
- mano specifico
- mano specifico
- Monouso/riutilizzabile:
- monouso
- monouso
- monouso
- monouso
- monouso
- monouso
- monouso
- monouso
- Talcati/non talcati:
- Non talcati
- Non talcati
- Non talcati
- Non talcati
- Non talcati
- Non talcati
- Non talcati
- Non talcati
- Classe di cleanroom:
- 10/ISO 4; EU GMP (A)
- 10/ISO 4; EU GMP (A)
- 10/ISO 4; EU GMP (A)
- 10/ISO 4; EU GMP (A)
- 10/ISO 4; EU GMP (A)
- 10/ISO 4; EU GMP (A)
- 10/ISO 4; EU GMP (A)
- 10/ISO 4; EU GMP (A)
- Tipo polsino:
- Arrotolato
- Arrotolato
- Arrotolato
- Arrotolato
- Arrotolato
- Arrotolato
- Cuff_Style-21391103|Cuff_Style-21391099
- Arrotolato
- Spessore polsino:
- 0,10 mm (3,94 mil)
- 0,10 mm (3,94 mil)
- 0,10 mm (3,94 mil)
- 0,10 mm (3,94 mil)
- 0,10 mm (3,94 mil)
- 0,10 mm (3,94 mil)
- 0,10 mm (3,94 mil)
- 0,10 mm (3,94 mil)
- Spessore delle dita:
- 0,21 mm (8.26 mil)
- 0,21 mm (8.26 mil)
- 0,21 mm (8.26 mil)
- 0,21 mm (8.26 mil)
- 0,21 mm (8.26 mil)
- 0,21 mm (8.26 mil)
- 0,21 mm (8.26 mil)
- 0,21 mm (8.26 mil)
- Spessore palmo:
- 0,18 mm (7,09 mil)
- 0,18 mm (7,09 mil)
- 0,18 mm (7,09 mil)
- 0,18 mm (7,09 mil)
- 0,18 mm (7,09 mil)
- 0,18 mm (7,09 mil)
- 0,18 mm (7,09 mil)
- 0,18 mm (7,09 mil)
- Norma EN:
- EN 455, EN 374 Type B
- EN 455, EN 374 Type B
- EN 455, EN 374 Type B
- EN 455, EN 374 Type B
- EN 455, EN 374 Type B
- EN 455, EN 374 Type B
- EN 455, EN 374 Type B
- EN 455, EN 374 Type B
Frequently Bought Together