BOD bottles, with robotic stopper, WHEATON®
BOD bottle, with glass robotic stopper, 128×272 mm (Ø×H), 2 L
Codice catalogo WHEA227580
Nome del fornitore: DWK Life Sciences
BOD bottles, with robotic stopper, WHEATON®
Codice catalogo WHEA227580
Nome del fornitore: DWK Life Sciences
Vai a
Product Details & Documents
Made of Type I borosilicate glass.
- With glass Robotic stopper
- Large capacity
We recommend this 2 Liter BOD Bottle for long-term BOD and ultimate BOD analysis.
Specifiche
- Capacità:2 L
- Descrizione:BOD bottle, with glass robotic stopper, 128×272 mm (Ø×H)