Orders LinkContactUsLinkComponent
Syringes with valves
Syringes with valves
Certificates
Syringes with valves
Syringes with valves
10 Options Available Below
Product Details & Documents
These syringes are pre-fitted with valves. They are ideal for sample storage and transportation or to pressurise samples prior to injection.
Product Family Options
Pre-fitted with removable needle and valve
Product Information
  • Descrizione
    Volume
    Tipo
    Availability
    Price
  • Syringe pre-fitted with removable needle and valve, PTFE tipped plunger
    0,5 ml
    500R-V-GT
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 205.00
    SFr. 205.00
    1 * 1 ST
       SFr. 205.00
  • Syringe pre-fitted with removable needle and valve, PTFE tipped plunger
    1 ml
    1MR-V-GT
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 225.00
    SFr. 225.00
    1 * 1 ST
       SFr. 225.00
  • Syringe pre-fitted with removable needle and valve, PTFE tipped plunger
    2,5 ml
    2.5MDR-V-GT
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 236.00
    SFr. 236.00
    1 * 1 ST
       SFr. 236.00
Pre-fitted with Luer lock valve
Product Information
  • Descrizione
    Volume
    Tipo
    Availability
    Price
  • Syringe pre-fitted with Luer lock valve, PTFE tipped plunger
    1 ml
    1MR-VLL-GT
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 218.00
    SFr. 218.00
    1 * 1 ST
       SFr. 218.00
  • Syringe pre-fitted with Luer lock valve, PTFE tipped plunger
    2,5 ml
    2.5MDR-VLL-GT
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 236.00
    SFr. 236.00
    1 * 1 ST
       SFr. 236.00
  • Syringe pre-fitted with Luer lock valve, PTFE tipped plunger
    5 ml
    5MDR-VLLMA-GT
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 236.00
    SFr. 236.00
    1 * 1 ST
       SFr. 236.00
  • Syringe pre-fitted with Luer lock valve, PTFE tipped plunger
    10 ml
    10MDR-VLLMA-GT
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 227.00
    SFr. 227.00
    1 * 1 ST
       SFr. 227.00
  • Syringe pre-fitted with Luer lock valve, PTFE tipped plunger
    25 ml
    25MDR-VLLMA-GT
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 422.00
    SFr. 422.00
    1 * 1 ST
       SFr. 422.00
  • Syringe pre-fitted with Luer lock valve, PTFE tipped plunger
    50 ml
    50MR-VLLMA-GT
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 526.00
    SFr. 526.00
    1 * 1 ST
       SFr. 526.00
  • Syringe pre-fitted with Luer lock valve, PTFE tipped plunger
    100 ml
    100MR-VLLMA-GT
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 614.00
    SFr. 614.00
    1 * 1 ST
       SFr. 614.00
Valve needles
Product Information
  • Catalog #
    Gauge
    Ø est
    Lunghezza
    Tipo punta
    Descrizione
    Availability
    Price
  • 23
    0,63 mm
    50 mm
    Side hole/dome
    Valve needle for syringes with valves
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 87.80
    SFr. 87.80
    1 * 1 ST
       SFr. 87.80
Product Information
  • Catalog #
    Gauge
    Ø est
    Lunghezza
    Tipo punta
    Descrizione
    Availability
    Price
  • 23
    0,63 mm
    50 mm
    Cone
    Valve needle for 25 µl - 2,5 ml syringes with valves
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 50.70
    SFr. 50.70
    1 * 1 ST
       SFr. 50.70
Syringe valves
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • Syringe valve VLL25/2.5M, push-pull with Luer lock, 25 µl - 2,5 ml syringe volume
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 140.00
    SFr. 140.00
    1 * 1 ST
       SFr. 140.00
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • Syringe valve VLLMA, push button with Luer lock, 5 ml - 2 l syringe volume
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 120.00
    SFr. 120.00
    1 * 1 ST
       SFr. 120.00
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • Syringe valve SLLV, Luer lock and luer tip, 50 µl - 2 l syringe volume
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 120.00
    SFr. 120.00
    1 * 1 ST
       SFr. 120.00
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • Syringe valve V25/2.5M-0.63C, push-pull with needle, 25 µl - 2,5 ml syringe volume
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 140.00
    SFr. 140.00
    1 * 1 ST
       SFr. 140.00