Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
Siringhe
Siringhe per cromatografia
Syringes for HPLC autosamplers, Spark Holland instruments
Syringes for HPLC autosamplers, Spark Holland instruments
548-1646
:
Trajan Scientific and Medical
Syringes for HPLC autosamplers, Spark Holland instruments
548-1646
:
Trajan Scientific and Medical
Per:
Spark Holland Promis LC autosampler
Tipo:
250D-SPARK-GT
Volume:
250 µl
Descrizione:
Gas-tight, with PTFE-tipped plunger, 1/4-28 UNF front thread
Seq:
01
Conf:
1