Orders LinkContactUsLinkComponent
Vario couplings
Vario couplings
Nome del fornitore:   Bohlender
Vario couplings
Nome del fornitore:  Bohlender
Certificates
Vario couplings
8 varianti
Product Details & Documents
Vario 2-part coupling to connect elastic tubing with non-elastic tubing made of PTFE, glass, metal or other plastics.
  • The elastic tubing is pushed on a cone and fixed on the coupling by means of a connecting nut
  • These couplings are available in two different versions, according to male and female threads to connect
Product Family Options
With Male thread
Informazioni sul prodotto
  • Ø foro
    Per
    Filettatura
    Ø int.tubo
    Disponibilità
    Price
  • BOHLD681-08
    Couplings with GL male thread
    Couplings with GL male thread
    0,8 mm
    Connecting laboratory screw joints and elastic tubing wall thickness 0,8 and 1,6 mm
    GL 14
    0,8 mm
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 80.20
    SFr. 80.20
    1 * 1 ST
       SFr. 80.20
  • BOHLD681-24
    Couplings with GL male thread
    Couplings with GL male thread
    3,2 mm
    Connecting laboratory screw joints and elastic tubing wall thickness 1,6 mm
    GL 14
    3,2 mm
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 75.10
    SFr. 75.10
    1 * 1 ST
       SFr. 75.10
  • BOHLD681-32
    Couplings with GL male thread
    Couplings with GL male thread
    4,0 mm
    Connecting laboratory screw joints and elastic tubing wall thickness 1,6 mm
    GL 14
    4,0 / 4,8 mm
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 76.90
    SFr. 76.90
    1 * 1 ST
       SFr. 76.90
  • BOHLD681-16
    Couplings with GL male thread
    Couplings with GL male thread
    1,6 mm
    Connecting laboratory screw joints and elastic tubing wall thickness 0,8 and 1,6 mm
    GL 14
    1,6 mm
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 79.00
    SFr. 79.00
    1 * 1 ST
       SFr. 79.00
  • BOHLD681-40
    Couplings with GL male thread
    Couplings with GL male thread
    6,4 mm
    Connecting laboratory screw joints and elastic tubing wall thickness 1,6 mm
    GL 18
    6,4 mm
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 127.00
    SFr. 127.00
    1 * 1 ST
       SFr. 127.00
  • BOHLD681-48
    Couplings with GL male thread
    Couplings with GL male thread
    8,0 mm
    Connecting laboratory screw joints and elastic tubing wall thickness 1,6 and 2,4 mm
    GL 18
    8 / 9,5 mm
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 174.00
    SFr. 174.00
    1 * 1 ST
       SFr. 174.00
With Female thread
Informazioni sul prodotto
  • Ø foro
    Per
    Filettatura
    Ø int.tubo
    Disponibilità
    Price
  • BOHLF778-08
    Couplings with UNF1/4"28G female thread
    Couplings with UNF1/4"28G female thread
    0,8 mm
    Connecting flanged PTFE
    elastic tubing with end fittings and wall thickness 0,8 and 1,6 mm
    1/4“ 28G UNF
    0,8 mm
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 63.60
    SFr. 63.60
    1 * 1 ST
       SFr. 63.60
  • BOHLF778-16
    Couplings with UNF1/4"28G female thread
    Couplings with UNF1/4"28G female thread
    1,6 mm
    Connecting flanged PTFE
    elastic tubing with end fittings and wall thickness 0,8 and 1,6 mm
    1/4“ 28G UNF
    1,6 mm
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 62.70
    SFr. 62.70
    1 * 1 ST
       SFr. 62.70