Strisce reattive, magnesio, Reflectoquant®, Supelco®
Catalog # 1.16124.0001
Supplier: Merck
CAS Number:
Specifications
- Metodo chimico:Phthalein complexone
- Conf:50 Tests
- Descrizione:Magnesio
- Environmentally Preferable:
- M-Clarity quality level:MQ100
- Range di misura:5 - 100 mg/l Mg
Specifications
About this item
Strisce per l'uso su RQflex 10, RQflex 10 Plus e RQflex® 20.
- Fast analysis with reliable results
- Barcode calibration
- Low analysis costs
- Environmentally friendly