ContactUsLinkComponent
Strisce reattive, solfito, MQuant™, Supelco®
Strisce reattive, solfito, MQuant™, Supelco®
Codice catalogo 1.10013.0001
Nome del fornitore:  Merck
Numero CAS:  
undefined
Strisce reattive, solfito, MQuant™, Supelco®
Codice catalogo 1.10013.0001
Nome del fornitore:  Merck
Numero CAS:  
Specifiche
  • Metodo chimico:
    Nitroprussiato/Zn-esacianoferrato
  • Conf:
    100 Tests
  • Descrizione:
    Solfito
  • M-Clarity quality level:
    MQ100
  • Tipo:
    MQ
  • Range di misura:
    10-40-80-180-400 mg/l SO₃
Specifiche
Informazioni su questo articolo
MQuant® (MQ) test strips can be considered as a mobile laboratory on a few square millimeters of plastic strip. As a screening method they provide the user with a rapid overview of the situation, which helps in making the choice of what, and perhaps if, further analysis is required, thereby leading to potentially substantial savings in time and analytical resources. The strip is a biologically-degradable polyester foil and the low reagent content, in the reaction zones, assists problem-free waste disposal.
  • Easy to handle: just dip, wait and measure visually against colour scale
  • Safe: no auxiliary reagents necessary
  • Immediate results within 2 minutes enables fast decisions
  • Portable: can be used inside or outside the laboratory