Test strips and reagents for semi-quantitative determinations of phosphate in water analyses, QUANTOFIX®
Catalog # 705-0320
Supplier: MACHEREY-NAGEL
CAS Number:
Specifications
- Metodo chimico:Molybdenum blue
- Conf:100 Tests
- Descrizione:Phosphate
- Seq:35
- Range di misura:0 - 100 mg/l PO₄3-