ContactUsLinkComponent
Laboratorio compatto MColortest™, Supelco®
Laboratorio compatto MColortest™, Supelco®
Catalog # 1.11151.0001
Supplier:  Merck
CAS Number:  
undefined
Laboratorio compatto MColortest™, Supelco®
Catalog # 1.11151.0001
Supplier:  Merck
CAS Number:  
Specifications
  • Conf:
    1 SET
  • Descrizione:
    MColortest™ Laboratorio compatto per analisi dell'acqua
  • Seq:
    01
  • M-Clarity quality level:
    MQ100
Specifications
About this item
Merck has identified a range of applications where a particular combination of tests is required for field use. The Compact Laboratory range meets these requirements with a sturdy portable plastic case, which can be used as a work surface, packed with all the kits of equipment and reagents needed for the evaluation.
  • Compact
  • Suitable for standing and flowing water
  • Ideal for schools and Universities
  • All your requirements both kits and equipment packed in a portable case
Ordering information: Refill packs are available for all parameters
Delivery: Supplied with step by step operating instructions and guides