Orders LinkContactUsLinkComponent
Overalls, Proshield® 8 Proper, model CCF5
 :   DuPont
Overalls, Proshield® 8 Proper, model CCF5
 :  DuPont
Overalls, Proshield® 8 Proper, model CCF5
Overalls, Proshield® 8 Proper, model CCF5
ProShield® Proper overalls are made of a proprietary DuPont non woven PE fabric (55 g/m²) that makes them incredibly tough, lightweight and resistant to tears and punctures.
  • Mandarin collar, nylon zipper with flap
  • Two thigh pockets
  • Stitched seams
  • Wrist and back elastication for good fit, open ankles (not elasticated)
  • Antistatically treated for comfort (not acc. EN 1149-5)
  • Can be re-used several times and may also be washed up to seven times
The overalls are comfortable because the material allows the passage of air and water vapour. They are ideal workwear for do-it-yourself, general maintenance and cleaning, manufacturing and other non hazardous applications as well as a semi re-usable garment for visitors.
Colour: Grey
 : Cat. I
  • Catalog No:
  • DUPOCCF5SGY00/S
  • DUPOCCF5SGY00/M
  • DUPOCCF5SGY00/L
  • DUPOCCF5SGY00/XL
  • DUPOCCF5SGY00/XXL
  • Modello:
  • CCF5
  • CCF5
  • CCF5
  • CCF5
  • CCF5
  • Antistatico:
  • +
  • +
  • +
  • +
  • +
  • Applicazioni per abiti:
  • General Purpose|Food
  • General Purpose|Food
  • General Purpose|Food
  • General Purpose|Food
  • General Purpose|Food
  • Peso:
  • 55 g/m²
  • 55 g/m²
  • 55 g/m²
  • 55 g/m²
  • 55 g/m²
Product Family Options
  • Colore
    Taglia dell'abbigliamento
  • grigio
    S
    1 * 100 ST
    1 * 100 ST - SFr. 1’560.00
    SFr. 1’560.00
    1 * 100 ST
       SFr. 1’560.00
    Specifications
    Cat. No.
    DUPOCCF5SGY00/S
    Modello
    CCF5
    Antistatico
    +
    Applicazioni per abiti
    General Purpose|Food
    Peso
    55 g/m²
  • grigio
    M
    1 * 100 ST
    1 * 100 ST - SFr. 1’560.00
    SFr. 1’560.00
    1 * 100 ST
       SFr. 1’560.00
    Specifications
    Cat. No.
    DUPOCCF5SGY00/M
    Modello
    CCF5
    Antistatico
    +
    Applicazioni per abiti
    General Purpose|Food
    Peso
    55 g/m²
  • grigio
    L
    1 * 100 ST
    1 * 100 ST - SFr. 1’560.00
    SFr. 1’560.00
    1 * 100 ST
       SFr. 1’560.00
    Specifications
    Cat. No.
    DUPOCCF5SGY00/L
    Modello
    CCF5
    Antistatico
    +
    Applicazioni per abiti
    General Purpose|Food
    Peso
    55 g/m²
  • grigio
    XL
    1 * 100 ST
    1 * 100 ST - SFr. 1’560.00
    SFr. 1’560.00
    1 * 100 ST
       SFr. 1’560.00
    Specifications
    Cat. No.
    DUPOCCF5SGY00/XL
    Modello
    CCF5
    Antistatico
    +
    Applicazioni per abiti
    General Purpose|Food
    Peso
    55 g/m²
  • grigio
    XXL
    1 * 100 ST
    1 * 100 ST - SFr. 1’560.00
    SFr. 1’560.00
    1 * 100 ST
       SFr. 1’560.00
    Specifications
    Cat. No.
    DUPOCCF5SGY00/XXL
    Modello
    CCF5
    Antistatico
    +
    Applicazioni per abiti
    General Purpose|Food
    Peso
    55 g/m²