Burette and thermometer clamp with shaft, AUTOMATIC
Catalog # BOCH5624
Supplier: BOCHEM
Specifications
- Per:Burette e termometri
- Larghezza di serraggio:0 - 25 mm
- Conf:1
Specifications
About this item
This chromium plated clamp with shaft (length 115 mm) is used for holding burettes or thermometer (Ø 0 - 25 mm).