Orders LinkContactUsLinkComponent
2-Adamantanone
Glass bottle for solids, 25 g
  MOLE23794765-25G
Marketsource
 :   Molekula
2-Adamantanone
  MOLE23794765-25G
 :  Molekula
Marketsource
 :  700-58-3
2-Adamantanone
Glass bottle for solids, 25 g
 :  700-58-3
1 * 25 g
 :  Adamantan-2-one
  •  :700-58-3
  •  :C₁₀H₁₄O
  •  :MFCD00074737
  •  :150.22 g/mol
  •  :228 °C (1013 hPa)
  •  :43 °C
  •  :1 g/cm³ (20 °C)
  •  :Ambiente
  •  :211-847-2
  • Tipo di conf.:
    Glass bottle for solids
  • Conf:
    25 g