Dropping bottle with head stopper
100 ml
Codice catalogo MARI3610105
Nome del fornitore: MARIENFELD
Vai a
Product Details & Documents
Glass, amber.
- Good chemical and physical properties
- For use in applications that do not involve high thermal stresses
- Amber coloured to protect light sensitive substanzes
Dropping bottle with flat head stopper and round shoulder.
Specifiche
- Capacità:100 ml