Staining jars acc. to Schiefferdecker
Staining jar, Schiefferdecker, 89×70,5×46 mm
Codice catalogo 631-0699
Nome del fornitore: MARIENFELD
Vai a
Product Details & Documents
Made of soda-lime glass, with cover.
These staining trough hold up to 10 slides (76×26 mm), or 10 pairs of slides placed adjacently back-to-back.
Jars made of heavy-walled soda lime glass with a relatively high coefficient of expansion, and it should not exposed to rapid temperature changes, to avoid cracks caused by tensions.
Specifiche
- Descrizione:Staining jar, Schiefferdecker
- L×P×A:89×70,5×46 mm