Orders LinkContactUsLinkComponent
Flasks, round bottom, with threaded GL side necks, BOLA
Flasks, round bottom, with threaded GL side necks, BOLA
Supplier:   Bohlender
Flasks, round bottom, with threaded GL side necks, BOLA
Supplier:  Bohlender
Certificates
Flasks, round bottom, with threaded GL side necks, BOLA
Flasks, round bottom, with threaded GL side necks, BOLA
2 Options Available Below
Product Details & Documents
PFA, transparent.
  • Very good chemical resistance
  • Excellent thermal resistance from −200 to +250 °C
  • Vacuum: suitable
Available with central ground joint neck and lateral screw threads GL 18.
Product Family Options
Product Information
  • Capacità
    Collo centrale
    Altezza
    Ø est
    Availability
    Price
  • 500 ml
    29 NS
    177 mm
    107 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 1’010.00
    SFr. 1’010.00
    1 * 1 ST
       SFr. 1’010.00
  • 250 ml
    29 NS
    149 mm
    88 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 865.00
    SFr. 865.00
    1 * 1 ST
       SFr. 865.00